missing translation for 'onlineSavingsMsg'
Learn More

Diethyl cyclopropane-1,1-dicarboxylate

Catalog No. AAH5656814
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5656814 25 g
AAH5656803 1 g
AAH5656806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5656814 Supplier Thermo Scientific Chemicals Supplier No. H5656814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1559-02-0 | C9H14O4 | 186.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1559-02-0
Molecular Formula C9H14O4
Molecular Weight (g/mol) 186.21
MDL Number MFCD00001283
InChI Key KYYUCZOHNYSLFV-UHFFFAOYSA-N
Synonym diethyl 1,1-cyclopropanedicarboxylate, 1,1-diethyl cyclopropane-1,1-dicarboxylate, 1,1-cyclopropanedicarboxylic acid diethyl ester, 1,1-cyclopropanedicarboxylic acid, diethyl ester, cyclopropane-1,1-dicarboxylic acid diethyl ester, 1,1-bis ethoxycarbonyl cyclopropane, diethylcyclopropan-1,1-dicarboxylat, diethyl-1,1-cyclopropanedicarboxylate, acmc-209dcw, ksc495e2n
PubChem CID 73790
IUPAC Name diethyl cyclopropane-1,1-dicarboxylate
SMILES CCOC(=O)C1(CC1)C(=O)OCC

Specifications

Density 1.055
Boiling Point 94°C to 96°C (10 mmHg)
Flash Point 87°C (189°F)
Odor Odorless
Refractive Index 1.433
Quantity 25 g
Beilstein 2049509
Formula Weight 186.21
Chemical Name or Material Diethyl cyclopropane-1,1-dicarboxylate
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 216-321-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only