missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Chlorothiophosphate 97.0+%, TCI America™
Specifications
Specifications
| Boiling Point | 51°C |
| Quantity | 500 g |
| UN Number | 2751 |
| Synonym | O,O′-Diethyl Phosphorochloridothioate, O,O′-Diethyl Phosphorothiochloridate, O,O′-Diethylthiophosphoryl Chloride |
| Formula Weight | 188.61 |
| Percent Purity | ≥97.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | Diethyl Chlorothiophosphate |
Safety and Handling
| EINECSNumber | (2)-2939 |
| RTECSNumber | TD1780000 |
| TSCA | Yes |