missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Chlorophosphate 97.0+%, TCI America™

Catalog No. D220625G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D220625G 25 g
D2206250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D220625G Supplier TCI America Supplier No. D220625G

Chemical Identifiers

CAS 814-49-3
Molecular Formula C4H10ClO3P
Molecular Weight (g/mol) 172.545
MDL Number MFCD00009075
InChI Key LGTLXDJOAJDFLR-UHFFFAOYSA-N
Synonym diethyl chlorophosphate, diethyl chlorophosphonate, diethyl phosphorochloridate, diethyl phosphorochloride, diethoxyphosphoryl chloride, o,o-diethyl chlorophosphate, diethoxyphosphorus oxychloride, phosphorochloridic acid, diethyl ester, o,o-diethyl chloridophosphate, o,o-diethyl chlorophosphonate
PubChem CID 13139
IUPAC Name 1-[chloro(ethoxy)phosphoryl]oxyethane
SMILES CCOP(=O)(OCC)Cl

Specifications

Color Yellow
Boiling Point 93°C
Quantity 25 g
UN Number 2927
Formula Weight 172.54
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Diethyl Chlorophosphate
RTECSNumber TD1400000
TSCA Yes