missing translation for 'onlineSavingsMsg'
Learn More

Diethyl chlorophosphate, 97+%

Catalog No. AAH3382022
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3382022 100 g
AAH3382014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3382022 Supplier Thermo Scientific Chemicals Supplier No. H3382022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 814-49-3 | C4H10ClO3P | 172.545 g/mol

A phosphorous halide used for biochemical research. Also used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A phosphorous halide used for biochemical research. Also used as an intermediate in organic synthesis.

Solubility
Soluble in alcohol and chloroform. Slightly soluble in water.

Notes
Moisture Sensitive. Store away from water/moisture. Store away from oxidizing agent.

Chemical Identifiers

CAS 814-49-3
Molecular Formula C4H10ClO3P
Molecular Weight (g/mol) 172.545
MDL Number MFCD00009075
InChI Key LGTLXDJOAJDFLR-UHFFFAOYSA-N
Synonym diethyl chlorophosphate, diethyl chlorophosphonate, diethyl phosphorochloridate, diethyl phosphorochloride, diethoxyphosphoryl chloride, o,o-diethyl chlorophosphate, diethoxyphosphorus oxychloride, phosphorochloridic acid, diethyl ester, o,o-diethyl chloridophosphate, o,o-diethyl chlorophosphonate
PubChem CID 13139
IUPAC Name 1-[chloro(ethoxy)phosphoryl]oxyethane
SMILES CCOP(=O)(OCC)Cl

Specifications

Density 1.194
Boiling Point 60°C (2 mmHg)
Flash Point 61°C (142°F)
Quantity 100 g
UN Number UN3278
Sensitivity Moisture sensitive
Solubility Information Soluble in alcohol and chloroform. Slightly soluble in water.
Formula Weight 172.55
Percent Purity ≥97%
Chemical Name or Material Diethyl chlorophosphate
Hazard Category H300+H310+H330-H314
Hazard Statement GHS H Statement
H300-H310-H330-H227
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Combustible liquid.
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, LIQUID, TOXIC, N.O.S.
EINECSNumber 212-369-4
RTECSNumber TD1400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only