missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diéthyle benzylmalonate 98,0+%, TCI America™
Spécifications
Spécifications
| Color | Incolore |
| Boiling Point | 300°C |
| Quantity | 25 mL |
| Formula Weight | 250.29 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquide |
| Chemical Name or Material | Diethyl Benzylmalonate |
Sécurité et manipulation
| TSCA | Yes |