missing translation for 'onlineSavingsMsg'
Learn More

Diethyl allyl(3-phenyl-2-propynyl)malonate, 96%, Thermo Scientific Chemicals

Catalog No. AAH2770203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2770203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2770203 Supplier Thermo Scientific Chemicals Supplier No. H2770203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 143633-91-4 | Molecular Formula: C19H22O4 | Molecular Weight: 314.381 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 143633-91-4
Molecular Formula C19H22O4
Molecular Weight (g/mol) 314.381
MDL Number MFCD09953493
InChI Key ZSPBIVGRCZUELN-UHFFFAOYSA-N
Synonym diethyl allyl 3-phenyl-2-propynyl malonate, diethyl 2-allyl-2-3-phenylprop-2-yn-1-yl malonate, propanedioic acid,2-3-phenyl-2-propyn-1-yl-2-2-propen-1-yl-, 1,3-diethyl ester, 1,3-diethyl 2-3-phenylprop-2-yn-1-yl-2-prop-2-en-1-yl propanedioate, acmc-20ed36, 3-phenylpropargylallylmalonic acid diethyl ester, allyl 3-phenyl-2-propynyl malonic acid diethyl ester, diethyl 1-phenyl-6-heptene-1-yne-4,4-dicarboxylate, diethyl 3-phenylprop-2-yn-1-yl prop-2-en-1-yl propanedioate
PubChem CID 10086992
IUPAC Name diethyl 2-(3-phenylprop-2-ynyl)-2-prop-2-enylpropanedioate
SMILES CCOC(=O)C(CC=C)(CC#CC1=CC=CC=C1)C(=O)OCC

Specifications

Refractive Index 1.514
Quantity 1 g
Formula Weight 314.38
Percent Purity 96%
Chemical Name or Material Diethyl allyl(3-phenyl-2-propynyl)malonate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only