missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Adipate 99.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | -20°C |
| Color | Colorless |
| Boiling Point | 251°C |
| Quantity | 500 mL |
| Formula Weight | 202.25 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Diethyl Adipate |
Sécurité et manipulation
| EINECSNumber | (2)-0861&(2)-0879 |
| RTECSNumber | AV1100000 |
| TSCA | Yes |