missing translation for 'onlineSavingsMsg'
Learn More

Diéthyle (4-Nitrobenzyl)phosphonate 97,0+%, TCI America™

Numéro de catalogue. D52085G
Modifier l'affichage
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
D52085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. D52085G Fournisseur TCI America Code fournisseur D52085G

Identifiants chimiques

CAS 2609-49-6
Molecular Formula C11H16NO5P
Molecular Weight (g/mol) 273.225
MDL Number MFCD00047747
InChI Key FORMFFDDQMCTCT-UHFFFAOYSA-N
Synonym diethyl 4-nitrobenzylphosphonate, diethyl 4-nitrobenzyl phosphonate, diethyl 4-nitrophenyl methylphosphonate, 1-diethoxyphosphorylmethyl-4-nitro-benzene, 1-diethoxyphosphorylmethyl-4-nitrobenzene, phosphonic acid, 4-nitrophenyl methyl-, diethyl ester, phosphonic acid, p-4-nitrophenyl methyl-, diethyl ester, zlchem 660, diethyl4-nitrobenzylphosphonate
PubChem CID 256065
IUPAC Name 1-(diéthyphosphorylméthylle)-4-nitrobenzène
SMILES CCOP(=O)(CC1=CC=C(C=C1)[N+](=O)[O-])OCC

Spécifications

Boiling Point 153°C
Quantity 5 g
Formula Weight 273.22
Percent Purity ≥97.0% (GC)
Physical Form Liquide
Chemical Name or Material Diethyl (4-Nitrobenzyl)phosphonate
EINECSNumber (3)-4097
TSCA No