missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 3-Hydroxyglutarate 95.0+%, TCI America™

Catalog No. D31965G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D31965G 5 g
D319625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D31965G Supplier TCI America Supplier No. D31965G
Only null left

Chemical Identifiers

CAS 32328-03-3
Molecular Formula C9H16O5
Molecular Weight (g/mol) 204.222
InChI Key OLLQYIBTJXUEEX-UHFFFAOYSA-N
Synonym 3-Hydroxyglutaric Acid Diethyl Ester
PubChem CID 96259
IUPAC Name diethyl 3-hydroxypentanedioate
SMILES CCOC(=O)CC(CC(=O)OCC)O

Specifications

Color Yellow
Boiling Point 107°C
Quantity 5 g
Formula Weight 204.22
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl 3-Hydroxyglutarate