missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (3-Bromopropyl)phosphonate 95.0+%, TCI America™

Numéro de catalogue. p-7128276
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
D44341G 1 g
D44345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. D44341G Fournisseur TCI America Code fournisseur D44341G
Il en reste null

Identifiants chimiques

CAS 1186-10-3
Molecular Formula C7H16BrO3P
Molecular Weight (g/mol) 259.08
MDL Number MFCD00800966
InChI Key PEIKTSJIUKYDPC-UHFFFAOYSA-N
Synonym (3-Bromopropyl)phosphonic Acid Diethyl Ester
PubChem CID 283500
IUPAC Name diethyl (3-bromopropyl)phosphonate
SMILES CCOP(=O)(CCCBr)OCC

Spécifications

Color Yellow
Boiling Point 97°C
Quantity 1 g
Formula Weight 259.08
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl (3-Bromopropyl)phosphonate