missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl (3-Bromopropyl)phosphonate 95.0+%, TCI America™
Spécifications
Spécifications
| Color | Yellow |
| Boiling Point | 97°C |
| Quantity | 5 g |
| Formula Weight | 259.08 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Diethyl (3-Bromopropyl)phosphonate |
Sécurité et manipulation
| TSCA | No |