missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 2-bromoethylphosphonate, 97%

Catalog No. AAB2115606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2115606 5 g
AAB2115614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2115606 Supplier Thermo Scientific Chemicals Supplier No. B2115606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5324-30-1 | C6H14BrO3P | 245.05 g/mol

It is employed as a reactant involved in radical coupling, synthesis of organosoluble zirconium phosphonate nanocomposites, click-chemistry, asymmetric epoxidation of unfunctionalized olefins, Negishi alkyl-aryl cross-coupling, and in synthesis of hydrolytically stable phosphonic acids for use as dental adhesives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a reactant involved in radical coupling, synthesis of organosoluble zirconium phosphonate nanocomposites, click-chemistry, asymmetric epoxidation of unfunctionalized olefins, Negishi alkyl-aryl cross-coupling, and in synthesis of hydrolytically stable phosphonic acids for use as dental adhesives.

Solubility
Soluble in water(20 g/L) (25°C).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 5324-30-1
Molecular Formula C6H14BrO3P
Molecular Weight (g/mol) 245.05
MDL Number MFCD00000239
InChI Key PINITSMLVXAASM-UHFFFAOYSA-N
Synonym diethyl 2-bromoethyl phosphonate, diethyl 2-bromoethylphosphonate, phosphonic acid, 2-bromoethyl-, diethyl ester, 2-bromoethyl phosphonic acid diethyl ester, o,o-diethyl 2-bromoethyl phosphonate, diethyl bromoethylphosphonate, diethyl-2-bromoethyl-phosphonate, pinitsmlvxaasm-uhfffaoysa, diethyl-2-bromoethyl phosphonate
PubChem CID 79218
IUPAC Name 1-bromo-2-diethoxyphosphorylethane
SMILES CCOP(=O)(CCBr)OCC

Specifications

Density 1.275
Boiling Point 98°C to 100°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4605
Quantity 5 g
Beilstein 1766077
Solubility Information Soluble in water(20g/L) (25°C).
Formula Weight 245.06
Percent Purity 97%
Chemical Name or Material Diethyl 2-bromoethylphosphonate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 226-193-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only