missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 2,5-Dibromoadipate 98.0+%, TCI America™

Catalog No. D39655G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39655G 5 g
D396525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39655G Supplier TCI America Supplier No. D39655G
Only null left

Chemical Identifiers

CAS 869-10-3
Molecular Formula C10H16Br2O4
Molecular Weight (g/mol) 360.04
MDL Number MFCD00075379
InChI Key UBCNJHBDCUBIPB-UHFFFAOYNA-N
Synonym diethyl 2,5-dibromoadipate, diethyl meso-2,5-dibromoadipate, 1,6-diethyl 2,5-dibromohexanedioate, 2,5-dibromoadipic acid diethyl ester, hexanedioic acid, 2,5-dibromo-, diethyl ester, diethyl2,5-dibromohexanedioate, 2,5-dibromohexanedioic acid diethyl ester, acmc-1bjoi, diethylmeso-dibromoadipate, diethylmeso-2,5-dibromoadipate
PubChem CID 241588
IUPAC Name 1,6-diethyl 2,5-dibromohexanedioate
SMILES CCOC(=O)C(Br)CCC(Br)C(=O)OCC

Specifications

Melting Point 66°C
Color White-Yellow
Boiling Point 134°C
Quantity 5 g
UN Number 3261
Formula Weight 360.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diethyl 2,5-Dibromoadipate
EINECSNumber (2)-4013
TSCA No