missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 2,2-Diethoxyethylphosphonate, TCI America™

Catalog No. D24235G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24235G 5 g
D242325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D24235G Supplier TCI America Supplier No. D24235G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7598-61-0
Molecular Formula C10H23O5P
Molecular Weight (g/mol) 254.263
MDL Number MFCD00009236
InChI Key LUQYELQXRPNKRY-UHFFFAOYSA-N
PubChem CID 82071
IUPAC Name 2-diethoxyphosphoryl-1,1-diethoxyethane
SMILES CCOC(CP(=O)(OCC)OCC)OCC

Specifications

Color Yellow
Boiling Point 130°C
Quantity 5 g
Synonym diethyl 2,2-diethoxyethylphosphonate, diethyl 2,2-diethoxyethyl phosphonate, diethyl2,2-diethoxyethylphosphonate, 2,2-diethoxyethylphosphonic acid diethyl ester, phosphonic acid, 2,2-diethoxyethyl-, diethyl ester, 2,2-diethoxyethyl diethoxyphosphino-1-one, diethyl phosphonacetaldehyde diethyl acetal, diethylphosphonoacetaldehyde diethylacetal, 2-diethoxyphosphoryl-1,1-diethoxy-ethane
Formula Weight 254.26
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl 2,2-Diethoxyethylphosphonate
TSCA No
Recommended Storage Refrigerator