missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 1-tetradecylphosphonate, 97+%

Catalog No. AAB2142614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2142614 25 g
AAB2142606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2142614 Supplier Thermo Scientific Chemicals Supplier No. B2142614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5191-09-3 | C8H8N2 | 132.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5191-09-3
MDL Number MFCD00015597
InChI Key CPWIYYOBPHXJIA-UHFFFAOYSA-N
PubChem CID 640154
SMILES CCCCCCCCCCCCCCP(=O)(OCC)OCC

Specifications

Melting Point 8°C
Density 0.922
Boiling Point 199°C to 201°C (3 mmHg)
Molecular Formula C18H39O3P
Refractive Index 1.4445
Linear Formula CH3(CH2)13P(O)(OCH2CH3)2
Quantity 25 g
Synonym diethyl 1-tetradecanephosphonate, diethyl 1-tetradecylphosphonate, diethyl tetradecylphosphonate, acmc-20al9h, diethyl tetradecane-1-phosphonate, tetradecylphosphonic acid diethyl ester, tetradecyl-phosphonic acid diethyl ester, 1-tetradecanephosphonic acid diethyl ester, phosphonic acid, tetradecyl-, diethyl ester
IUPAC Name 1-diethoxyphosphoryltetradecane
Molecular Weight (g/mol) 334.48
Formula Weight 334.47
Percent Purity 98%
Chemical Name or Material Diethyl 1-tetradecylphosphonate
Show More Show Less
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only