missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 1-tetradecylphosphonate, 97+%

Catalog No. AAB2142606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2142606 5 g
AAB2142614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2142606 Supplier Thermo Scientific Chemicals Supplier No. B2142606
Only null left

CAS: 5191-09-3 | C8H8N2 | 132.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5191-09-3
MDL Number MFCD00015597
InChI Key CPWIYYOBPHXJIA-UHFFFAOYSA-N
PubChem CID 640154
ChEBI CHEBI:33068
SMILES CCCCCCCCCCCCCCP(=O)(OCC)OCC

Specifications

Melting Point 8°C
Density 0.922
Boiling Point 199°C to 201°C (3 mmHg)
Molecular Formula C8H8N2
Linear Formula CH3(CH2)13P(O)(OCH2CH3)2
Refractive Index 1.4445
Quantity 5 g
Synonym 7-aminoindole, 7-amino-1h-indole, 1h-indol-7-ylamine, 7-indolamine, indole-7-ylamine, 7-indolylamine, 7-aminoindol, acmc-1ay6v, indol-7-yl amine, 7-aminoindole hplc
IUPAC Name 1H-indol-7-amine
Molecular Weight (g/mol) 132.17
Formula Weight 334.47
Percent Purity 98%
Chemical Name or Material Diethyl 1-tetradecylphosphonate
Show More Show Less
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only