missing translation for 'onlineSavingsMsg'
Learn More

Diethyl 1-decylphosphonate, 97%

Catalog No. AAL1303814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1303814 25 g
AAL1303806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1303814 Supplier Thermo Scientific Chemicals Supplier No. L1303814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16165-68-7 | C14H31O3P | 278.373 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16165-68-7
Molecular Formula C14H31O3P
Molecular Weight (g/mol) 278.373
MDL Number MFCD00015593
InChI Key HYQXIVAZQBYBAF-UHFFFAOYSA-N
Synonym diethyl 1-decylphosphonate, diethyl decylphosphonate, decylphosphonic acid diethyl ester, decyldiethoxyphosphino-1-one, acmc-20al9l, diethyl n-decanephosphonate, diethyl 1-decanephosphonate, diethyl 1-decyl phosphonate, decyl-phosphonic acid diethyl ester, n-decylphosphonic acid diethyl ester
PubChem CID 2773697
IUPAC Name 1-diethoxyphosphoryldecane
SMILES CCCCCCCCCCP(=O)(OCC)OCC

Specifications

Boiling Point 128°C to 130°C (0.2 mmHg)
Refractive Index 1.439
Quantity 25 g
Beilstein 1784261
Formula Weight 278.38
Percent Purity 97%
Chemical Name or Material Diethyl 1-decylphosphonate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only