missing translation for 'onlineSavingsMsg'
Learn More

Diethoxymethylsilane 95.0+%, TCI America™

Catalog No. D240325ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D240325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D240325ML Supplier TCI America Supplier No. D240325ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2031-62-1
Molecular Formula C5H13O2Si
Molecular Weight (g/mol) 133.242
MDL Number MFCD00026749
InChI Key GAURFLBIDLSLQU-UHFFFAOYSA-N
Synonym diethoxymethylsilane, methyldiethoxysilane, diethoxy methyl silane, silane, diethoxymethyl, diethoxy-methylsilane, diethoxy methyl silicon, diethoxymethylsilane. methylhydrogendiethoxysilane., diethoxymethyl-silan, diethoxy methyl silyl
PubChem CID 6327387
IUPAC Name diethoxy(methyl)silicon
SMILES CCO[Si](C)OCC

Specifications

Color Yellow
Boiling Point 97°C
Quantity 25 mL
UN Number 1993
Formula Weight 134.25
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethoxymethylsilane [Hydrosilylating Reagent]
EINECSNumber (2)-2052&(2)-2053
TSCA Yes