missing translation for 'onlineSavingsMsg'
Learn More

Diethoxy(methyl)phenylsilane 97.0+%, TCI America™

Catalog No. D376025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D376025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D376025G Supplier TCI America Supplier No. D376025G

Chemical Identifiers

CAS 775-56-4
Molecular Formula C11H18O2Si
Molecular Weight (g/mol) 210.35
MDL Number MFCD00048541
InChI Key MNFGEHQPOWJJBH-UHFFFAOYSA-N
Synonym methylphenyldiethoxysilane, diethoxy methyl phenylsilane, diethoxy methyl phenyl silane, silane, diethoxymethylphenyl, benzene, diethoxymethylsilyl, pubchem10811, acmc-209pap, methyl-phenyl-diethoxysilane, diethoxy-methyl-phenyl-silane
PubChem CID 69891
IUPAC Name diethoxy(methyl)phenylsilane
SMILES CCO[Si](C)(OCC)C1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 218°C
Quantity 25 g
Formula Weight 210.35
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethoxy(methyl)phenylsilane