Learn More
Diesel Fuel #2 Composite Standard, Restek™ DFS Item
Supplier:  Chromatographic RK31259
 View more versions of this product
Catalog No. 06-801-433
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
Diesel Fuel #2 Composite Standard
Methylene Chloride
50,000μg/mL
Ampule
5 mL
Product Suggestions
Product Suggestions