missing translation for 'onlineSavingsMsg'
Learn More

Dienestrol 96.0+%, TCI America™

Catalog No. D0449100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0449100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D0449100MG Supplier TCI America Supplier No. D0449100MG

Chemical Identifiers

CAS 84-17-3
Molecular Formula C18H18O2
Molecular Weight (g/mol) 266.34
MDL Number MFCD00050983
InChI Key NFDFQCUYFHCNBW-SCGPFSFSSA-N
PubChem CID 667476
IUPAC Name 4-[(2E,4E)-4-(4-hydroxyphenyl)hexa-2,4-dien-3-yl]phenol
SMILES CC=C(C1=CC=C(C=C1)O)C(=CC)C2=CC=C(C=C2)O

Specifications

Melting Point 229°C
Quantity 100 mg
Formula Weight 266.34
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dienestrol
RTECSNumber SL0580000
TSCA No