missing translation for 'onlineSavingsMsg'
Learn More

Didodecyldimethylammonium Chloride 98.0+%, TCI America™

Catalog No. D46985G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D46985G Supplier TCI America Supplier No. D46985G

Chemical Identifiers

CAS 3401-74-9
Molecular Formula C26H56ClN
Molecular Weight (g/mol) 418.191
MDL Number MFCD00038724
InChI Key WLCFKPHMRNPAFZ-UHFFFAOYSA-M
Synonym Dilauryldimethylammonium Chloride
PubChem CID 18843
IUPAC Name didodecyl(dimethyl)azanium;chloride
SMILES CCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCC.[Cl-]

Specifications

Melting Point 131°C
Color White-Yellow
Quantity 5 g
Formula Weight 418.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Didodecyldimethylammonium Chloride
EINECSNumber (2)-0184&(9)-1971
RTECSNumber JR2120000
TSCA Yes