missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Didecyl Disulfide 95.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 208°C |
| Quantity | 10 mL |
| Formula Weight | 346.68 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Didecyl Disulfide |
Safety and Handling
| EINECSNumber | (2)-0473&(2)-0475 |
| TSCA | No |