missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dicycloverine Hydrochloride, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| Chemical Name or Material | Pharmacopeia & Metrological Institutes Standards |
| CAS | 67-92-5 |
| Flash Point | Not Applicable |
| Molecular Formula | C19H35NO2 · HCl |
| Linear Formula | C19H35NO2 · HCl |
| Quantity | 100 mg |
| MDL Number | MFCD00079158 |
| Synonym | Dicyclomine hydrochloride; 2-(Diethylamino)ethyl 1-cyclohexylcyclohexane-1-carboxylate hydrochloride; [1,1-Bicyclohexyl]-1-carboxylic acid 2-(diethylamino)ethyl ester hydrochloride |
| Formula Weight | 345.95 |
| Grade | BP |