missing translation for 'onlineSavingsMsg'
Learn More

Dicyandiamide 98.0+%, TCI America™

Catalog No. C0454500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0454500G 500 g
C045425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0454500G Supplier TCI America Supplier No. C0454500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 461-58-5
Molecular Formula C2H4N4
Molecular Weight (g/mol) 84.08
MDL Number MFCD00008066
InChI Key QGBSISYHAICWAH-UHFFFAOYSA-N
Synonym dicyandiamide, cyanoguanidine, dicyanodiamide, 1-cyanoguanidine, n-cyanoguanidine, guanidine, cyano, pyroset do, dicyandiamido, epicure dicy 7, dicyandiamin
PubChem CID 10005
IUPAC Name N''-cyanoguanidine
SMILES NC(N)=NC#N

Specifications

Melting Point 211°C
Color White
Quantity 500 g
Formula Weight 84.08
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dicyandiamide
EINECSNumber (2)-1694
RTECSNumber ME9950000
TSCA Yes