missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichlorotitanium Diisopropoxide 95.0+%, TCI America™
Spécifications
Spécifications
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 236.94 |
| Percent Purity | ≥95.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Dichlorotitanium Diisopropoxide |
Sécurité et manipulation
| TSCA | Yes |
| Recommended Storage | Freezer |