missing translation for 'onlineSavingsMsg'
Learn More

Dichlorophenylsilane, 96%

Catalog No. AAL0437114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0437114 25 g
AAL0437106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0437114 Supplier Thermo Scientific Chemicals Supplier No. L0437114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1631-84-1 | C6H5Cl2Si | 176.09 g/mol

Dichlorophenylsilane is used as the intermediates of OLED,pharmaceuticals, cosmetics. It is also used as reduction reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dichlorophenylsilane is used as the intermediates of OLED,pharmaceuticals, cosmetics. It is also used as reduction reagent.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Store away from moisture, oxidizing agents. Incompatible materials are bases, moisture and oxidizing agents.

Chemical Identifiers

CAS 1631-84-1
Molecular Formula C6H5Cl2Si
Molecular Weight (g/mol) 176.09
MDL Number MFCD00039296
InChI Key XNAFLNBULDHNJS-UHFFFAOYSA-N
Synonym phenyldichlorosilane, dichlorophenylsilane, dichloro phenyl silane, dichloro phenyl silicon, benzene, dichlorosilyl, phenyl dichlorosilane, phenyl hydrogen dichlorosilane, phenyl-dichlorosilane, phenyldichloro silane, dichloro phenyl silyl
PubChem CID 6327324
SMILES Cl[Si](Cl)C1=CC=CC=C1

Specifications

Boiling Point 181°C
Quantity 25 g
UN Number UN2986
Beilstein 2433865
Flash Point 48°C (118°F)
Solubility Information Hydrolyzes in water.
IUPAC Name dichloro(phenyl)silyl
Formula Weight 177.11
Percent Purity 96%
Refractive Index 1.523
Sensitivity Moisture sensitive
Density 1.212
Chemical Name or Material Dichlorophenylsilane
Show More Show Less
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, FLAMMABLE, N.O.S.
EINECSNumber 216-635-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only