missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethylenedimethyliminium Chloride 97.0+%, TCI America™

Catalog No. D15975G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D15975G 5 g
D159725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D15975G Supplier TCI America Supplier No. D15975G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 33842-02-3
Molecular Formula C3H6Cl3N
Molecular Weight (g/mol) 162.44
MDL Number MFCD00011870
InChI Key NRNFKRFWZQQDMD-UHFFFAOYSA-M
Synonym dichloromethylene dimethylammonium chloride, phosgeniminium chloride, n-dichloromethylene-n-methylmethanaminium chloride, methanaminium, n-dichloromethylene-n-methyl-, chloride, dichloromethylene-dimethyliminium chloride, dichloromethylenedimethylammonium chloride, dichloromethylidene dimethylazanium chloride, phosgeneiminium chloride, phosgene-iminium chloride
PubChem CID 2723945
IUPAC Name dichloro(dimethylamino)methylium chloride
SMILES [Cl-].CN(C)[C+](Cl)Cl

Specifications

Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 162.44
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dichloromethylenedimethyliminium Chloride