missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethyl Methyl Ether 97.0+%, TCI America™

Catalog No. D164525G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D164525G 25 g
D1645250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D164525G Supplier TCI America Supplier No. D164525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4885-02-3
Molecular Formula C2H4Cl2O
Molecular Weight (g/mol) 114.95
MDL Number MFCD00000828
InChI Key GRTGGSXWHGKRSB-UHFFFAOYSA-N
Synonym dichloromethyl methyl ether, 1,1-dichlorodimethyl ether, dichloro methoxy methane, methane, dichloromethoxy, ether, dichloromethyl methyl, alpha,alpha-dichloromethyl methyl ether, methoxydichloromethane, 1,1-dichloromethyl methyl ether, alpha,alpha-dichloromethyl ether, dichloromethylmethylether
PubChem CID 21004
IUPAC Name dichloro(methoxy)methane
SMILES COC(Cl)Cl

Specifications

Color Yellow
Boiling Point 85°C
Quantity 25 g
UN Number 2924
Formula Weight 114.95
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Dichloromethyl Methyl Ether
RTECSNumber KN8150000
TSCA Yes