missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, CHROMASOLV™ Plus, for HPLC, contains 50-150 ppm amylene as stabilizer, ≥99.9%, Solstice

Catalog No. 60048238
Change view
Click to view available options
Quantity:
4 x 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60048238 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60048238 Supplier Solstice Supplier No. 6504634X4L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CHROMASOLV™ Plus, for HPLC, contains 50-150 ppm amylene as stabilizer,≥99.9%

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -97°C (literature)
Absorbance max. 0.10 at 240 nm, max. 1.00 at 235 nm, max. 0.005 at 400 nm, max. 0.005 at 300 nm, max. 0.02 at 250 nm
Density 1.325 g/mL at 25 °C(lit.)
Quantity 4 x 4 L
Packaging Glass Bottle
Linear Formula CH2Cl2
Boiling Point 39.8°C to 40°C (literature)
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material Dichloromethane, CHROMASOLV™ Plus, for HPLC, contains 50-150 ppm amylene as stabilizer, ≥99.9%
ShelfLife 547 days from date of manufacture
EINECSNumber 200-838-9