missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, 99.8%, for spectroscopy, stabilized with amylene

CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol

Supplier:  Thermo Scientific Chemicals 167770025

 View more versions of this product

Catalog No. AC167770025



Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
Specifications

Specifications

Dichloromethane
Stabilized
(1 cm cell vs HPLC−grade water), (1 cm cell vs HPLC−grade water) at 0nm, 0.01 max. at 270nm, 0.01 max. at 280nm, 0.02 max. at 260nm, 0.05 max. at 250nm, 0.2 max. at 240nm, 1 max. at 233nm
39.0°C to 40.0°C
Glass Bottle
0.0003% max.
1.325
CH2Cl2
0.43 mPa.s (20°C)
01, 60
15, 6135
Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf
ClCCl
84.93
CHEBI:15767
99.8%
-97.0°C
75-09-2 , 513-35-9
1.3250g/mL
2.5 L
Liquid
99.8%
CH2Cl2
MFCD00000881
0.02% max.
01,676; 02,273; 04,337; 07,239; 13,106
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
YMWUJEATGCHHMB-UHFFFAOYSA-N
dichloromethane
6344
84.93
Spectroscopy

Chemical Identifiers

75-09-2
84.93
YMWUJEATGCHHMB-UHFFFAOYSA-N
6344
dichloromethane
CH2Cl2
MFCD00000881
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
CHEBI:15767
ClCCl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May cause drowsiness or dizziness. Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure.

GHS P Statement Use personal protective equipment as required. Obtain special instructions before use. Do not handle until all safety precautions have been read and understood. Avoid breathing dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If exposed or you feel unwell: Call a POISON CENTRE or doctor/physician.

Warning

EINECSNumber : 200-838-9

RTECSNumber : PA8050000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only