Learn More
Dichloromethane, 99.8%, for spectroscopy, stabilized with amylene
CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
$100.89 - $281.00
Chemical Identifiers
| CAS | 75-09-2 |
|---|---|
| Molecular Formula | CH2Cl2 |
| Molecular Weight (g/mol) | 84.93 |
| MDL Number | MFCD00000881 |
| InChI Key | YMWUJEATGCHHMB-UHFFFAOYSA-N |
| Synonym | methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm |
| PubChem CID | 6344 |
| ChEBI | CHEBI:15767 |
| IUPAC Name | dichloromethane |
| SMILES | ClCCl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC167775000
|
Thermo Scientific Chemicals
167775000 |
500 mL | Glass Bottle |
Each for $100.89
Case of 6 Each for $0.00
|
|
||||
|
AC167770025
|
Thermo Scientific Chemicals
167770025 |
2.5 L | Glass Bottle |
Each for $281.00
Case of 4 Each for $0.00
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopyChemical Identifiers
| 75-09-2 | |
| 84.93 | |
| YMWUJEATGCHHMB-UHFFFAOYSA-N | |
| 6344 | |
| dichloromethane |
| CH2Cl2 | |
| MFCD00000881 | |
| methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
| CHEBI:15767 | |
| ClCCl |
Specifications
| -97.0°C | |
| (1 cm cell vs HPLC−grade water), (1 cm cell vs HPLC−grade water) at 0nm, 0.01 max. at 270nm, 0.01 max. at 280nm, 0.02 max. at 260nm, 0.05 max. at 250nm, 0.2 max. at 240nm, 1 max. at 233nm | |
| 39.0°C to 40.0°C | |
| Glass Bottle | |
| 0.0003% max. | |
| 1.325 | |
| CH2Cl2 | |
| 0.43 mPa.s (20°C) | |
| 01, 60 | |
| 15, 6135 | |
| Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf | |
| ClCCl | |
| 84.93 | |
| CHEBI:15767 | |
| 99.8% | |
| Dichloromethane, Stabilized |
| 75-09-2,513-35-9 | |
| 1.3250g/mL | |
| 500 mL | |
| Liquid | |
| 99.8% | |
| CH2Cl2 | |
| MFCD00000881 | |
| 0.02% max. | |
| 01,676; 02,273; 04,337; 07,239; 13,106 | |
| methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
| YMWUJEATGCHHMB-UHFFFAOYSA-N | |
| dichloromethane | |
| 6344 | |
| 84.93 | |
| Spectroscopy |
Safety and Handling
GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May cause drowsiness or dizziness. Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure.
GHS P Statement Use personal protective equipment as required. Obtain special instructions before use. Do not handle until all safety precautions have been read and understood. Avoid breathing dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If exposed or you feel unwell: Call a POISON CENTRE or doctor/physician.
Warning
EINECSNumber : 200-838-9
RTECSNumber : PA8050000
TSCA : TSCA
RUO – Research Use Only