CAS RN 513-35-9
2-Methyl-2-butene, 2M solution in THF, AcroSeal™
CAS: 513-35-9 | C5H10 | 70.14 g/mol
2-Methyl-2-butene, tech. 90%, remainder mainly 2-methyl-1-butene
CAS: 513-35-9 Molecular Formula: C5H10 Molecular Weight (g/mol): 70.14 MDL Number: MFCD00009276 InChI Key: BKOOMYPCSUNDGP-UHFFFAOYSA-N Synonym: 2-methyl-2-butene,2-butene, 2-methyl,trimethylethylene,amylene,n-amylene,3-methyl-2-butene,ethylene, trimethyl,beta-isoamylene,1,1,2-trimethylethylene,2-methylbutene-2 PubChem CID: 10553 ChEBI: CHEBI:77916 IUPAC Name: 2-methylbut-2-ene SMILES: CC=C(C)C
2-Methyl-2-butene, 90%, balance 2-Methyl-1-butene
CAS: 513-35-9 Molecular Formula: C5H10 Molecular Weight (g/mol): 70.14 MDL Number: MFCD00009276 InChI Key: BKOOMYPCSUNDGP-UHFFFAOYSA-N Synonym: 2-methyl-2-butene,2-butene, 2-methyl,trimethylethylene,amylene,n-amylene,3-methyl-2-butene,ethylene, trimethyl,beta-isoamylene,1,1,2-trimethylethylene,2-methylbutene-2 PubChem CID: 10553 ChEBI: CHEBI:77916 IUPAC Name: 2-methylbut-2-ene SMILES: CC=C(C)C
2-Methyl-2-butene, ≥99% (GC), MilliporeSigma™ Supelco™
2-Methyl-2-butene is a trisubstituted olefin which is mostly used as a precursor for peroxyacetyl nitrate. It can be prepared by the acid-catalyzed dehydration of 3-methyl-2-butanol. It has 3 alkyl groups on the double bond.
2-Methyl-2-butene, 99+%
CAS: 513-35-9 Molecular Formula: C5H10 Molecular Weight (g/mol): 70.14 MDL Number: MFCD00009276 InChI Key: BKOOMYPCSUNDGP-UHFFFAOYSA-N Synonym: 2-methyl-2-butene,2-butene, 2-methyl,trimethylethylene,amylene,n-amylene,3-methyl-2-butene,ethylene, trimethyl,beta-isoamylene,1,1,2-trimethylethylene,2-methylbutene-2 PubChem CID: 10553 ChEBI: CHEBI:77916 IUPAC Name: 2-methylbut-2-ene SMILES: CC=C(C)C
2-Methyl-2-butene 90.0+%, TCI America™
CAS: 513-35-9 Molecular Formula: C5H10 Molecular Weight (g/mol): 70.14 MDL Number: MFCD00009276 InChI Key: BKOOMYPCSUNDGP-UHFFFAOYSA-N Synonym: 2-methyl-2-butene,2-butene, 2-methyl,trimethylethylene,amylene,n-amylene,3-methyl-2-butene,ethylene, trimethyl,beta-isoamylene,1,1,2-trimethylethylene,2-methylbutene-2 PubChem CID: 10553 ChEBI: CHEBI:77916 IUPAC Name: 2-methylbut-2-ene SMILES: CC=C(C)C
2-Methyl-2-butene 95.0+%, TCI America™
CAS: 513-35-9 Molecular Formula: C5H10 Molecular Weight (g/mol): 70.14 MDL Number: MFCD00009276 InChI Key: BKOOMYPCSUNDGP-UHFFFAOYSA-N Synonym: 2-methyl-2-butene,2-butene, 2-methyl,trimethylethylene,amylene,n-amylene,3-methyl-2-butene,ethylene, trimethyl,beta-isoamylene,1,1,2-trimethylethylene,2-methylbutene-2 PubChem CID: 10553 ChEBI: CHEBI:77916 IUPAC Name: 2-methylbut-2-ene SMILES: CC=C(C)C