missing translation for 'onlineSavingsMsg'
Learn More

Dichloroacetonitrile 98.0+%, TCI America™

Catalog No. D2510500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2510500G 500 g
D251025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D2510500G Supplier TCI America Supplier No. D2510500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3018-12-0
Molecular Formula C2HCl2N
Molecular Weight (g/mol) 109.937
MDL Number MFCD00040886
InChI Key STZZWJCGRKXEFF-UHFFFAOYSA-N
Synonym dichloroacetonitrile, acetonitrile, dichloro, dichloromethyl cyanide, ccris 942, unii-o0l64v63m9, dichloro acetonitrile, pubchem17242, acmc-209hdn, dsstox_cid_1562
PubChem CID 18177
ChEBI CHEBI:82444
IUPAC Name 2,2-dichloroacetonitrile
SMILES C(#N)C(Cl)Cl

Specifications

Color Yellow
Boiling Point 112°C
Quantity 500 g
UN Number 2920
Formula Weight 109.94
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dichloroacetonitrile
RTECSNumber AL8465000
TSCA No