missing translation for 'onlineSavingsMsg'
Learn More

Dichloro(1,5-cyclooctadiene)palladium(II) 98.0+%, TCI America™

Catalog No. D26041G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26041G 1 g
D26045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26041G Supplier TCI America Supplier No. D26041G
Only null left

Chemical Identifiers

CAS 12107-56-1
Molecular Formula C8H12Cl2Pd
Molecular Weight (g/mol) 285.50
MDL Number MFCD00012412
InChI Key RRHPTXZOMDSKRS-PHFPKPIQSA-L
Synonym dichloro 1,5-cyclooctadiene palladium ii, cycloocta-1,5-diene dichloropalladium, 1,5-cyclooctadienepalladium ii dichloride, 1z,5z-cycloocta-1,5-diene; palladium 2+ ; dichloride, dichloro 1,5-cyclooctadiene palladium, palladium, dichloro 1,5-cyclooctadiene, dichloro 1,2,5,6-eta-cycloocta-1,5-diene palladium, palladium, dichloro 1,2,5,6-eta-1,5-cyclooctadiene, dichloro 1,5-cyclooctadiene palladium ii ,, dichloro 1,5-cyclooctadiene palladium ii 1g
PubChem CID 6436380
IUPAC Name palladium(2+) (1Z,5Z)-cycloocta-1,5-diene dichloride
SMILES [Cl-].[Cl-].[Pd++].C1C\C=C/CC\C=C/1

Specifications

Melting Point 210°C
Quantity 1 g
Formula Weight 285.50
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dichloro(1,5-cyclooctadiene)palladium(II)