missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloro[1,1'-bis(dicyclohexylphosphino)ferrocene]palladium(II), Pd 14.1%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04638303
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Dichloro[1,1'-bis(dicyclohexylphosphino)ferrocene]palladium(II) | |
| 294°C to 300°C | |
| C34H52Cl2FeP2Pd | |
| 1 g | |
| [Fe].Cl[Pd]Cl.C1CCC(CC1)P(C1CCCCC1)c1cccc1.C1CCC(CC1)P(C1CCCCC1)c1cccc1 | |
| 755.90 | |
| Pd 14.1% |
| 917511-90-1 | |
| Orange | |
| MFCD11656081 | |
| PdCl2(dcypf); [1,1'-Bis(dicyclohexylphosphino)ferrocene]dichloropalladium(II) | |
| Dichloro[1,1'-bis(dicyclohexylphosphino)ferrocene]palladium(II) | |
| 755.91 | |
| Crystalline Solid |
Chemical Identifiers
| 917511-90-1 | |
| 755.90 | |
| PdCl2(dcypf); [1,1'-Bis(dicyclohexylphosphino)ferrocene]dichloropalladium(II) | |
| [Fe].Cl[Pd]Cl.C1CCC(CC1)P(C1CCCCC1)c1cccc1.C1CCC(CC1)P(C1CCCCC1)c1cccc1 |
| C34H52Cl2FeP2Pd | |
| MFCD11656081 | |
| Dichloro[1,1'-bis(dicyclohexylphosphino)ferrocene]palladium(II) |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only