missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Dicer knockout Mouse embryonic stem cells Lysate
Description
The Dicer knockout Mouse embryonic stem cells Lysate is derived from mouse. It has been validated for the following applications: Western Blot, Immunoprecipitation.
Specifications
Specifications
| Quantity | 0.25 mg |
| Storage Buffer | Standard RIPA buffer including protease and phosphatase inhibitors and 0.1% SDS. No reducing agents or loading buffer added. |
| Purity | Protein |
| Cellular Fraction | Whole |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Product Line | Novus |
| For Use With (Application) | Immunoprecipitation, Western Blot |
| Tissue | Embryo |
For Research Use Only