missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl Phthalate 97.0+%, TCI America™

Catalog No. P0292500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0292500ML 500 mL
P029225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0292500ML Supplier TCI America Supplier No. P0292500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 84-74-2
Molecular Formula C16H22O4
Molecular Weight (g/mol) 278.35
MDL Number MFCD00009441
InChI Key DOIRQSBPFJWKBE-UHFFFAOYSA-N
Synonym dibutyl phthalate, di-n-butyl phthalate, n-butyl phthalate, butyl phthalate, celluflex dpb, elaol, genoplast b, palatinol c, polycizer dbp, unimoll db
PubChem CID 3026
ChEBI CHEBI:34687
IUPAC Name 1,2-dibutyl benzene-1,2-dicarboxylate
SMILES CCCCOC(=O)C1=CC=CC=C1C(=O)OCCCC

Specifications

Melting Point -35°C
Color Yellow
Boiling Point 340°C
Quantity 500 mL
UN Number 3082
Formula Weight 278.35
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Dibutyl Phthalate
EINECSNumber (3)-1303
RTECSNumber TI0875000
TSCA Yes