missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl phthalate, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111019234
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019234 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019234 Supplier Merck Emd Millipore Supplier No. 1828150MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.

Specifications

Packaging Bottomless Glass Bottle with Fused Cone
CAS 84-74-2
Product Line TraceCERT
Vapor Pressure 1 mmHg (147°C)
Vapor Density 9.6 (vs. air)
Density 1.043 g/mL (at 25°C)
Physical Form Neat
Linear Formula C6H4-1,2-[CO2(CH2)3 CH3]2
MDL Number MFCD00009441
Melting Point -35°C
Boiling Point 340°C
Synonym n-Butyl phthalate; DBP; Phthalic acid dibutyl ester
Quantity 50 mg
Formula Weight 278.34
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC