missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibutyl Malonate 99.0+%, TCI America™
Spécifications
Spécifications
| Color | Colorless |
| Boiling Point | 251°C |
| Quantity | 25 g |
| Formula Weight | 216.28 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Dibutyl Malonate |
Sécurité et manipulation
| TSCA | No |