missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibutyle Fumarate 98,0+%, TCI America™
Spécifications
Spécifications
| Melting Point | -18°C |
| Color | Incolore |
| Boiling Point | 285°C |
| Quantity | 500 g |
| Formula Weight | 228.29 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquide |
| Chemical Name or Material | Dibutyl Fumarate |
Sécurité et manipulation
| EINECSNumber | (2)-1099&(2)-1142 |
| RTECSNumber | LT1225000 |
| TSCA | Yes |