missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibucaine hydrochloride, 99%, MilliporeSigma™ Supelco™
Dibucaine hydrochloride is a quinolone derivative, which can be prepared using isatin as the starting material followed by a series of reactions. It is a local anesthetic agent.
Supplier: Merck Emd Millipore SGMAD063825G
Description
Featured Industries: Pharmaceutical (small molecule)Specifications
| 61-12-1 | |
| C20H29N3O2 · HCl | |
| 99°C to 101°C | |
| Analytical Standard | |
| 379.92 | |
| Analytical Standard |
| Neat | |
| MFCD00012735 | |
| 2-Butoxy-N-(2-diethylaminoethyl)-4-quinolinecarboxamide hydrochloride; Cinchocaine hydrochloride | |
| 25 g | |
| ≥99% |