CAS RN 61-12-1
IUPAC Name:
{2-[(2-butoxyquinolin-4-yl)formamido]ethyl}diethylazanium chloride
Synonyms:
dibucaine hydrochloride dibucaine hcl cinchocaine hcl cinchocaine hydrochloride
Molecular Weight (g/mol):
379.93
Molecular Formula:
C20H30ClN3O2
InChi Key:
IVHBBMHQKZBJEU-UHFFFAOYSA-N
SMILES:
[Cl-].CCCCOC1=CC(C(=O)NCC[NH+](CC)CC)=C2C=CC=CC2=N1
1
–
2
of
2
results
1
Dibucaine hydrochloride
CAS: 61-12-1 Molecular Formula: C20H30ClN3O2 Molecular Weight (g/mol): 379.929 MDL Number: MFCD00012735 InChI Key: IVHBBMHQKZBJEU-UHFFFAOYSA-N Synonym: dibucaine hydrochloride,dibucaine hcl,cinchocaine hydrochloride,cinchocaine hcl,cincaine chloride,dibucaine cinchocaine hcl,percain,percaine,sovcain PubChem CID: 521951 ChEBI: CHEBI:59735 IUPAC Name: 2-butoxy-N-[2-(diethylamino)ethyl]quinoline-4-carboxamide;hydrochloride SMILES: CCCCOC1=NC2=CC=CC=C2C(=C1)C(=O)NCCN(CC)CC.Cl
2
Dibucaine hydrochloride, 99%, MilliporeSigma™ Supelco™
Dibucaine hydrochloride is a quinolone derivative, which can be prepared using isatin as the starting material followed by a series of reactions. It is a local anesthetic agent.