missing translation for 'onlineSavingsMsg'
Learn More

Dibucaine hydrochloride, 99%, MilliporeSigma™ Supelco™

Catalog No. p-7187523
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014877 1 g
111014879 5 g
111014878 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111014877 Supplier Merck Emd Millipore Supplier No. D06381G
Only null left

Dibucaine hydrochloride is a quinolone derivative, which can be prepared using isatin as the starting material followed by a series of reactions. It is a local anesthetic agent.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 61-12-1
Physical Form Neat
Linear Formula C20H29N3O2 · HCl
MDL Number MFCD00012735
Melting Point 99°C to 101°C
Synonym 2-Butoxy-N-(2-diethylaminoethyl)-4-quinolinecarboxamide hydrochloride; Cinchocaine hydrochloride
Type Analytical Standard
Quantity 1 g
Formula Weight 379.92
Percent Purity ≥99%
Grade Analytical Standard
Show More Show Less