missing translation for 'onlineSavingsMsg'
Learn More

Dibromo(1,5-cyclooctadiene)palladium(II), Pd 28.4%

Catalog No. AA4649203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4649203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4649203 Supplier Thermo Scientific Chemicals Supplier No. 04649203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 12145-47-0 | C8H12Br2Pd | 374.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12145-47-0
Molecular Formula C8H12Br2Pd
Molecular Weight (g/mol) 374.41
MDL Number MFCD00799635
InChI Key MTEMVOIUTLIPJT-PHFPKPIQSA-L
Synonym dibromo 1,5-cyclooctadiene palladium ii, pdbr2 cod, dibromo 1,5-cyclooctadiene palladium, 1,5-cyclooctadiene, z,z-; palladium ii bromide
PubChem CID 11187970
IUPAC Name (1Z,5Z)-cycloocta-1,5-diene;dibromopalladium
SMILES Br[Pd+]Br.C1C\C=C/CC\C=C/1

Specifications

Melting Point 201°C to 205°C
Color Orange
Quantity 1 g
Formula Weight 374.41
Concentration or Composition (by Analyte or Components) Pd 28.4%
Physical Form Powder
Chemical Name or Material Dibromo(1,5-cyclooctadiene)palladium(II)
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only