missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl Sulfoxide 99.0+%, TCI America™

Catalog No. D000125G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D000125G 25 g
D0001250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D000125G Supplier TCI America Supplier No. D000125G

Chemical Identifiers

CAS 621-08-9
Molecular Formula C14H14OS
Molecular Weight (g/mol) 230.33
MDL Number MFCD00004782
InChI Key HTMQZWFSTJVJEQ-UHFFFAOYSA-N
Synonym dibenzyl sulfoxide, benzyl sulfoxide, dibenzylsulfoxide, tardiol d, sulfoxide, dibenzyl, dibenzyl sulphoxide, sulfinylbis methylene dibenzene, bis phenylmethyl sulfoxide, nsc 55, benzene, 1,1'-sulfinylbis methylene bis
PubChem CID 12116
IUPAC Name (phenylmethanesulfinylmethyl)benzene
SMILES O=S(CC1=CC=CC=C1)CC1=CC=CC=C1

Specifications

Melting Point 133°C
Color White-Yellow
Quantity 25 g
Formula Weight 230.33
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Dibenzyl Sulfoxide
RTECSNumber DA9275000
TSCA Yes