missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl Phosphite 95.0+%, TCI America™

Catalog No. P101625G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P101625G 25 g
P1016250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P101625G Supplier TCI America Supplier No. P101625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17176-77-1
Molecular Formula C14H14O3P
Molecular Weight (g/mol) 261.24
MDL Number MFCD00004774
InChI Key RQKYHDHLEMEVDR-UHFFFAOYSA-N
Synonym dibenzyl phosphonate, phosphonic acid dibenzyl ester, unii-1o720l5h5a, phosphonic acid, bis phenylmethyl ester, dibenzylphosphit, dibenzylphosphonate, phosphonic acid bis phenylmethyl ester, oxo-bis phenylmethoxy phosphanium, mibxhgzaarwagi-uhfffaoysa-n, dibenzyl phosphite, technical grade
PubChem CID 6334615
IUPAC Name bis(benzyloxy)(oxo)-λ⁵-phosphanylium
SMILES O=[P+](OCC1=CC=CC=C1)OCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 165°C
Quantity 25 g
Formula Weight 262.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dibenzyl Phosphite
EINECSNumber (3)-3484
TSCA No