missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl Disulfide 98.0+%, TCI America™

Catalog No. B041725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B041725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B041725G Supplier TCI America Supplier No. B041725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 150-60-7
Molecular Formula C14H14S2
Molecular Weight (g/mol) 246.386
MDL Number MFCD00004783
InChI Key GVPWHKZIJBODOX-UHFFFAOYSA-N
Synonym dibenzyl disulfide, benzyl disulfide, disulfide, bis phenylmethyl, benzyl bisulfide, 1,2-dibenzyldisulfane, dibenzyl disulphide, dibenzyldisulfid, bis phenylmethyl disulfide, di phenylmethyl disulfide, disulfide, dibenzyl
PubChem CID 9012
ChEBI CHEBI:72752
IUPAC Name (benzyldisulfanyl)methylbenzene
SMILES C1=CC=C(C=C1)CSSCC2=CC=CC=C2

Specifications

Melting Point 72°C
Color White-Yellow
Quantity 25 g
Formula Weight 246.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dibenzyl Disulfide
EINECSNumber (3)-1136
RTECSNumber JO1750000
TSCA Yes