missing translation for 'onlineSavingsMsg'
Learn More

Dibenzoylmethane, 98%

Catalog No. AC112605000
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC112605000 500 g Plastic bottle
AC112600100 10 g Glass Bottle
AC112600250 25 g Plastic bottle
AC112601000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC112605000 Supplier Thermo Scientific Chemicals Supplier No. 112605000

CAS: 120-46-7 | C15H12O2 | 224.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 120-46-7
Molecular Formula C15H12O2
Molecular Weight (g/mol) 224.26
MDL Number MFCD00003085
InChI Key NZZIMKJIVMHWJC-UHFFFAOYSA-N
Synonym dibenzoylmethane, 1,3-diphenyl-1,3-propanedione, 2-benzoylacetophenone, phenyl phenacyl ketone, 1,3-propanedione, 1,3-diphenyl, rhodiastab 83, omega-benzoylacetophenone, dibenzoyl-methane, karenzu dk2, unii-ans7me8okc
PubChem CID 8433
ChEBI CHEBI:75417
IUPAC Name 1,3-diphenylpropane-1,3-dione
SMILES C1=CC=C(C=C1)C(=O)CC(=O)C2=CC=CC=C2

Specifications

Melting Point 75°C to 79°C
Color Yellow
Boiling Point 219°C to 221°C (18.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula C6H5COCH2COC6H5
Quantity 500 g
Beilstein 07,769
Merck Index 15,3014
Formula Weight 224.26
Percent Purity 98%
Physical Form Crystals
Chemical Name or Material Dibenzoylmethane, 98%
Show More Show Less
EINECSNumber 204-398-9
RTECSNumber TZ1930000
TSCA TSCA

RUO – Research Use Only