missing translation for 'onlineSavingsMsg'
Learn More

Dibenzoyl peroxide, 75%, remainder water

Catalog No. p-64202
Change view
Click to view available options
Quantity:
5 g
100 g
1 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC211780050 5 g Plastic bottle
AC211781000 100 g Plastic bottle
AC211780010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC211780050 Supplier Thermo Scientific Chemicals Supplier No. 211780050
Only null left

CAS: 94-36-0 | C14H10O4 | 242.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Radical initiators

Chemical Identifiers

CAS 94-36-0 , 7732-18-5
Molecular Formula C14H10O4
Molecular Weight (g/mol) 242.23
MDL Number MFCD00003071
InChI Key OMPJBNCRMGITSC-UHFFFAOYSA-N
Synonym benzoyl peroxide, dibenzoyl peroxide, peroxide, dibenzoyl, benzoperoxide, benzoyl superoxide, acetoxyl, lucidol, benoxyl, panoxyl, benzol peroxide
PubChem CID 7187
ChEBI CHEBI:82405
IUPAC Name benzoyl benzenecarboperoxoate
SMILES C1=CC=C(C=C1)C(=O)OOC(=O)C2=CC=CC=C2

Specifications

Melting Point 104.0°C to 106.0°C
Color White
Packaging Plastic bottle
Linear Formula (C6H5CO)2O2
Quantity 5 g
Beilstein 09, 179
Fieser 01,196; 04,122; 05,182; 06,160; 07,89; 12,157; 15,146; 17,367
Merck Index 15, 1119
Solubility Information Solubility in water: insoluble. Other solubilities: sparingly soluble in alcohol,soluble in benzene,chloroform,ether and acetone,1g/40 mL carbon disulfide,1g/40 mL olive oil
Formula Weight 242.23
Percent Purity 75%
Physical Form Wet Granular Powder
Chemical Name or Material Dibenzoyl peroxide, 75%, remainder water
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Causes serious eye irritation.
Heating may cause a fire.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Keep/Store away from clothing/ combustible materials.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash
EINECSNumber 202-327-6
RTECSNumber DM8575000
TSCA TSCA

RUO – Research Use Only